missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50, Thermo Scientific Chemicals
321.34 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black-Gray to Gray |
Melting Point | 1350.0°C |
Density | 3.4600g/mL |
Quantity | 2.5 kg |
Fieser | 01,723; 02,293; 05,570; 06,502; 07,312; 01,723; 02,293; 05,570; 06,502; 07,312; 08,433; 09,405; 10,339; 11,457; 12,422; 13,265; 14,270; 15,278; 17,296 |
Formula Weight | 85.68 |
Packaging | Glass bottle |
Form | Powder |
Chemical Name or Material | Aluminum-nickel |
Safety and Handling
Product Identifier
- Aluminium-nickel, Raney-type alloy, powder
- Danger
- Carcinogenicity Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H351-Suspected of causing cancer.
- H261-In contact with water releases flammable gases.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'