Learn More
Aluminium nitrate nonahydrate, 99+%, for analysis, ACROS Organics™
Brand: Acros Organics 218280025
Code : 29
Additional Details : CAS Number : 7784-27-2 Weight : 2.50000kg
Packaging | Glass bottle |
---|---|
Quantity | 2.5kg |
Chemical Identifiers
7784-27-2 | |
375.13 | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
aluminum;trinitrate;nonahydrate |
AlN3O9·9H2O | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
24567 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
Specifications
Aluminium nitrate nonahydrate | |
99% min. (Complexometry) | |
Al(NO3)3·9H2O | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate | |
24567 | |
Crystals | |
Analysis | |
20ppm max. | |
20ppm max. | |
Solubility in water: 64 g/100mL (25°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ethyl acetate and, pyridine | |
Undesignated | |
2.5 to 3.5 (5% soln. at 20°C) |
7784-27-2 | |
AlN3O9·9H2O | |
15,347 | |
Solubility in water: 64g/100mL (25°C). Other solubilities: soluble in alcohol,slightly soluble in acetone,practically insoluble in ethyl acetate and,pyridine | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
375.13 | |
375.13 | |
99+% | |
20ppm max | |
20ppm max. | |
Glass bottle | |
50ppm max. | |
73.0°C | |
2.5kg |
Safety and Handling
- Aluminium nitrate nonahydrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.