missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Basic, Brockmann Activity Grade I, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5390670
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]

High quality chemicals for laboratory and specialized industrial use

    For Chromatography
Physical Form Powder
Quantity 500 g
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade BAKER ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.00kg
Chemical Name or Material Aluminum Oxide, Basic, Brockmann Activity Grade I
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'