missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum spheres, 7.94mm (0.31 in.) dia., 99% (metals basis)

Brand:  Alfa Aesar™ 042031.KP

Code : NEW

Additional Details : CAS Number : 7429-90-5 Weight : 0.01000kg

Quantity 500 pc.

 View more versions of this product

Product Code. 15476415

  • 186.00 € / Pack of 500

Estimated Shipment date
from Supplier 24-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00213972
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
CHEBI:33629
[Al]

Specifications

Alloy 1100, Al:Cu; ≥99:0.12 wt%
500 pc.
7429-90-5
Al
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
[Al]
26.982
CHEBI:33629
99%
Aluminum
7.94mm (0.31 in.) dia.
(metals basis)
MFCD00213972
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
5359268
Spheres
Videos
SDS