Learn More
Emma Safety Footwear Amazone Safety Shoes
On top of being an extremely safe shoe, Amazone also offers its wearer exceptional comfort. The steel toe cap will protect from falling objects, while the easy twist sole provides a 'sticky grip'.
Brand: Emma Safety Footwear MM33254646
Additional Details : Weight : 0.40000kg
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
46 |
Laces | |
Amazone | |
Oiled Nubuck | |
Steel |