missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Amber Wide Neck Glass Reagent Bottles with Flat Head Stopper

Brand:  Duran™ 211888606

Code : A0

Additional Details : Weight : 3.89000kg

 View more versions of this product

Product Code. 10008010

  • 404.00 € / Each


Estimated Shipment
24-02-2020
Add to basket

Specifications

Specifications

10 L
Amber
Standard Ground Joint, Glass Flat-Head Stopper
85/55
Reagent Bottle
443mm
ISO 4796-2, USP Standard
Suitable for storage of aggressive media, which could attack plastic parts.
Glass
10Pack
229mm
Wide Mouth
Videos
Documents