Learn More
Invitrogen™ Acrylamide/Bis 19:1, 40% (w/v) solution
Ambion Acrylamide/Bis 19:1 is a 40% (w/v) solution of acrylamide (38%) and bis-acrylamide (2%) ideal for use in ribonuclease protection assay, sequencing gels, and sizing DNA or RNA fragments.
218.00 € - 349.00 €
Specifications
Content And Storage | Store at 4°C. |
---|---|
Format | Bottle |
Form | Liquid |
Product Line | Ambion™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10298364
|
Invitrogen™
AM9022 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10695785
|
Invitrogen™
AM9024 |
2 x 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Ambion Acrylamide/Bis 19:1 is a 40% (w/v) solution of acrylamide (38%) and bis-acrylamide (2%) ideal for use in ribonuclease protection assay, sequencing gels, and sizing DNA or RNA fragments. The solution is provided in a ready-to-use form, reducing the dust, inhalation, and contact hazard associated with weighing and preparing acrylamide and bis-acrylamide powders and solutions. Concentrations are carefully controlled and all solutions are micro-filtered prior to packaging. All Ambion acrylamides are rigorously tested for contaminating non-specific endonuclease, exonuclease, and RNase activity, and for functionality.
Order Info
Shipping Condition: Room temperature
Specifications
Store at 4°C. | |
Bottle | |
Liquid | |
Ambion™ |
For Research Use Only. Not for use in diagnostic procedures.