Learn More
Invitrogen™ Yeast tRNA (5 mg/mL)
Purified Saccharomyces cerevisiae yeast RNA is is a very effective blocking agent when used in Northern prehybridization and hybridization buffers at a concentration of 100–200 μg/mL, as well as being suitable as a coprecipitant in nucleic acid precipitations.
Brand: Invitrogen™ AM7120G
132.05 EUR valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.10500kg
Description
Purified Saccharomyces cerevisiae yeast RNA is is a very effective blocking agent when used in Northern prehybridization and hybridization buffers at a concentration of 100–200 μg/mL, as well as being suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactions inhibited by exogenous RNA, and it is the most inexpensive source of a high-quality coprecipitant. This total RNA preparation is supplied in one tube containing 0.5 mL at a concentration of 5 mg/mL in diethyl pyrocarbonate-treated distilled water. It is typically used at a working concentration of 10–20 μg/mL.
What is a Coprecipitant?
Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.
Order Info
Shipping Condition: Dry Ice
Specifications
Store at –20°C. | |
Carrier for Nucleic Acid Precipitation | |
0.5 mL |
5 mg/mL | |
Yeast transfer RNA | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.