Learn More
Cytiva Amersham™ Hybond™-N Membranes
Use for high sensitivity applications in DNA and RNA blotting. Cytiva Life Sciences™Amersham™ Hybond™-N Membranes is a strong supported membrane that is inherently hydrophilic and requires no pre-wetting.
Description
- Binding capacity for nucleic acids up to 600μm/cm2
- Inherently hydrophilic and requires no prewetting
- Nucleic acids can be crosslinked using UV light
Specifications
| Up to 600μg/cm2 | |
| Nylon, Neutral |