Learn More
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Brand: Cytiva 10600094
111.27 EUR valid until 2024-12-31
Use promo code "21973" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 9004-70-0 Weight : 0.01000kg
Description
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Individual sheets.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
8 cm | |
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
Sheet |
9 cm | |
25/Pk. | |
0.2 μm |
Safety and Handling
- Amersham Protran
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.