Learn More
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Brand: Cytiva 10600073
107.52 EUR valid until 2024-12-31
Use promo code "21973" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00600kg
Description
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Individual sheets.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
10 cm | |
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
Sheet | |
0.45 μm | |
Amersham Protran |
10 cm | |
10/Pk. | |
Nitrocellulose | |
10 cm x 10 cm |
Safety and Handling
- Amersham Protran
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.