missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Ultra-4 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
153.00 € - 1165.00 €
Spécification
Disposable | Single-use |
---|---|
Material (Membrane) | High-recovery Ultracel-PL Regenerated Cellulose |
Sterility | Non-sterile |
Filtration Area | 3 cm2 |
For Use With (Application) | Protein purification, protein concentration and desalting |
Code produit | Marque | Diameter (Metric) | MWCO | Volume (Metric) | Quantity | Prix | Quantité et disponibilité | ||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Diameter (Metric) | MWCO | Volume (Metric) | Quantity | Prix | Quantité et disponibilité | ||
10131842
|
Merck
UFC805008 |
22.7 mm | 50 kDa | 4 mL | 8Pack |
154.00 €
Pack of 8 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
10191813
|
Merck
UFC800308 |
22.7 mm | 3 kDa | 4 mL | 8Pack |
154.00 €
Pack of 8 |
N/A | ||
Currently Unavailable: We have no information when this product will be available
Voir les produits de remplacement
|
10384823
|
Merck
UFC800324 |
17.3 mm | 3 kDa | 4 mL | 24Pack |
443.00 €
Pack of 24 |
In Stock
Log in to see stock. |
|
10283383
|
Merck
UFC800396 |
17.3 mm | 3 kDa | 4 mL | 96Pack |
1165.00 €
Pack of 96 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||
10640753
|
Merck
UFC801008 |
22.7 mm | 10 kDa | 4 mL | 8 Pack |
153.00 €
Pack of 8 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||
10341782
|
Merck
UFC801024 |
17.2 mm | 10 kDa | 4 mL | 24Pack |
409.00 €
Pack of 24 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||
10158911
|
Merck
UFC801096 |
17.3 mm | 10 kDa | 4 mL | 96 Pack |
1155.00 €
Pack of 96 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||
10677872
|
Merck
UFC803008 |
22.7 mm | 30 kDa | 4 mL | 8Pack |
153.00 €
Pack of 8 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
10687872
|
Merck
UFC803024 |
22.7 mm | 30 kDa | 4 mL | 24Pack |
366.00 €
Pack of 24 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
10254762
|
Merck
UFC805024 |
22.7 mm | 50 kDa | 4 mL | 24Pack |
472.00 €
Pack of 24 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
10540962
|
Merck
UFC810008 |
22.7 mm | 100 kDa | 4 mL | 8Pack |
154.00 €
Pack of 8 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
10638262
|
Merck
UFC810024 |
22.7 mm | 100 kDa | 4 mL | 24Pack |
416.00 €
Pack of 24 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||
10698062
|
Merck
UFC810096 |
22.7 mm | 100 kDa | 4 mL | 96Pack |
1160.00 €
Pack of 96 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
10210342
|
Merck
UFC803096 |
22.7 mm | 30 kDa | 4 mL | 96Pack |
1155.00 €
Pack of 96 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
10264762
|
Merck
UFC805096 |
22.7 mm | 50 kDa | 4 mL | 96Pack |
1160.00 €
Pack of 96 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
Description
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
- High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
- Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
- Heat-sealed membrane eliminates possibility of downstream extractables
- Universal rotor compatibility
- High retentate recovery of >90%
- High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
- Easy sample monitoring due to translucent housing and volume gradations
- Direct pipetter sample access eliminates processing step to recover concentrate
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates
- Concentration of dilute or prepurified proteins from column eluents
- Desalting and buffer exchange
Spécification
Single-use | |
Non-sterile | |
Protein purification, protein concentration and desalting | |
Two microcentrifuge tubes | |
4 mL |
High-recovery Ultracel-PL Regenerated Cellulose | |
3 cm2 | |
Regenerated Cellulose (RC) | |
Centrifugal Filters | |
124 mm |