Learn More
Technical Service Consultants™ Amies transport buffer clear prepared culture media sterile in bijou
Brand: Technical Service Consultants™ M06B008
Code : 03
Additional Details : Weight : 0.00800kg
Description
Ready prepared Amies transport buffer and Amies transport charcoal media.Recommended for transporting and preserving microbiological samples over 24 hours.Supplied in sterile 8mL bijou bottles.