missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoacetonitrile hydrochloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
Description
Used as pharmaceutical intermediates, organic synthetic raw material transportation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 166°C to 171°C (decomposition) |
CAS | 6011-14-9 |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | >98% |
Molecular Formula | C2H5ClN2 |
MDL Number | MFCD00012850 |
Synonym | aminoacetonitrile hydrochloride, 2-aminoacetonitrile hydrochloride, glycinonitrile hydrochloride, aminoacetonitrile hcl, unii-s8l3v1ksb7, acetonitrile, amino-, monohydrochloride, s8l3v1ksb7, aminoacetonitrile.hcl, amino-acetonitrile hcl, acmc-209mhz |
Solubility Information | Soluble in water 20°C,1000g/L. |
InChI Key | XFKYKTBPRBZDFG-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'