missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
195.34 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 125°C |
Color | White |
Ignition Residue | 0.3% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% |
Packaging | Plastic drum |
Linear Formula | H2NNHC(=NH)NH2·H2CO3 |
Quantity | 5 kg |
Beilstein | 03,117 |
Solubility Information | Solubility in water: <5g/L (20°C) |
Show More |
Safety and Handling
Product Identifier
- Aminoguanidine bicarbonate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Respiratory sensitiser Category 1
- H411-Toxic to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'