Learn More
WTW™ Ammonium Cell Test Kit
With certificate for optimum precision.
Brand:  WTW™ 250495
Code : A0
Additional Details : Weight : 0.25000kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Product Code. 11771043
  • 130.00 € / 0.25kg
Estimated Shipment: 13-05-2024
to see stock.
Description
Description
  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For infrequent or very large sample throughput in the laboratory
  • Sample adding, inserting, measuring and reading
  • Reaction cuvette with barcode or method selection
Specifications
Specifications
Indophenol blue (Berthelot reaction)
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers
25
25 Tests
CombiCheck Ammonium (NH4) cell test kit
For waste water, drinking water, sea water
Certificates
0.010 - 2.000mg/I
Photometric