missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, ca. 7N solution in methanol, ACROS Organics™

Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger

Brand:  Acros Organics 133710010

Code : X2

Additional Details : CAS Number : 7664-41-7 Weight : 0.82000kg Transport : UN number : 3286 Chem class : 3 Pack group : II


Packaging Glass Bottle
Quantity 1L

 View more versions of this product

Product Code. 10001310

  • 150.00 € / 1 L

In stock, ready to ship: 302

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger

Specifications

Chemical Identifiers

7664-41-7
17.03
QGZKDVFQNNGYKY-UHFFFAOYSA-N
222
azane
H3N
MFCD00011418
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
CHEBI:16134
N

Specifications

Ammonia
7664-41-7,67-56-1
H3N
0.7700g/mL
05,15
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
N
17.03
CHEBI:16134
Liquid
0.77
Colorless to Yellow
1L
ca. 7N Solution in Methanol
6.0 to 7.5N
NH3
MFCD00011418
15, 488
QGZKDVFQNNGYKY-UHFFFAOYSA-N
azane
222
17.03
Glass Bottle
0.5% max.
14°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonia, ca 7N solution in methanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

SDS Documents
Certificates