missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, 35%, for HPLC, Fisher Chemical™
32.86 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
100mL, Plastic Coated Glass bottle
Description
H5NO, CAS Number-1336-21-6, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid
Specifications
pH | 12 |
Color | Colorless |
CAS | 1336-21-6 |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
SMILES | N.O |
IUPAC Name | amine hydrate |
Molecular Weight (g/mol) | 35.05 |
Formula Weight | 35.05g/mol |
Show More |
Safety and Handling
Product Identifier
- Ammonia solution 35%
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Ammonium hydroxide,Ammonia
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'