missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide solution, Eluent additive for LC-MS, ≥25% in H2O, Honeywell Fluka™

Catalog Number 15628410
Click to view available options
Quantity:
100 mL
10 x 1 mL
Packaging:
Ampoule
Glass Bottle
Polybottle
1336-21-6, 7732-18-5
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
N.O
1336-21-6, 7732-18-5
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
N.O

Eluent additive for LC-MS,≥25% in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 24°C
CAS Min % ≥70.0000%, ≥20.0000%
CAS Max % <90.0000%, <30.0000%
Linear Formula NH4OH
UN Number UN2672
Beilstein 3587154
Density 0.9 g/mL at 25 °C(lit.)
Quantity 10 x 1 mL
IUPAC Name azanium;hydroxide
Explosion Limit 0.27
Formula Weight 35.05
Percent Purity >25%
Grade LC-MS
Packaging Ampoule
Chemical Name or Material Ammonium hydroxide solution, Eluent additive for LC-MS, ≥25% in H2O
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ammonia, aqueous solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'