missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(II) Sulfate Hexahydrate, Certified AR for analysis, Fisher Chemical™
55.48 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
1 kg
3 kg
Description
Ammonium Iron(II) Sulfate Hexahydrate, Certified AR for analysis
Specifications
Melting Point | 100°C |
Color | Green |
pH | 3,5 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 392.14g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Ammonium iron (II) sulfate hexahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'