missing translation for 'onlineSavingsMsg'
Learn More

Ammonium oxalate monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog Number p-59645
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
6009-70-7
C2H8N2O4·H2O
142.11
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
6009-70-7
C2H8N2O4·H2O
142.11
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.50%
Linear Formula (CO2NH4)2·H2O
Merck Index 15, 534
Solubility Information Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 142.11
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Ammonium oxalate monohydrate, p.a.
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'