missing translation for 'onlineSavingsMsg'
Learn More

Ammonium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™

Catalog Number p-7083600
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy
NH4+
This item is not returnable. View return policy
NH4+

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 1g/L
Packaging Bottle
Quantity 100 mL
Type Single-component Standard Solution
Solvent or Matrix H2O
Physical Form Liquid
Chemical Name or Material Ammonium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'