missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Fisher BioReagents

Catalog Number p-24429
37.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Poly Bottle
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
sulfuric acid diamine
N.N.OS(O)(=O)=O
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
sulfuric acid diamine
N.N.OS(O)(=O)=O
This enzyme-grade Ammonium Sulfate is suitable for use in the purification of proteins.
TRUSTED_SUSTAINABILITY
Melting Point 280°C
Color Colorless
pH 5 to 6
Physical Form Solid
Quantity 1 kg
Linear Formula (NH4)2SO4
Merck Index 15, 552
Formula Weight 132.14
Percent Purity ≥99.5%
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Ammonium Sulfate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'