missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiosulfate, 96%, pure, Thermo Scientific Chemicals

Catalog Number p-4089758
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
7783-18-8
H8N2O3S2
148.21
MFCD00015965
ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate
This item is not returnable. View return policy
7783-18-8
H8N2O3S2
148.21
MFCD00015965
ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 150.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 95% min. (Iodimetry)
Linear Formula (NH4)2S2O3
Merck Index 15, 559
Solubility Information Solubility in water: 63.1% (20°C) - 71.0% (70°C). Other solubilities: insoluble in alcohol and ether
Formula Weight 148.21
Percent Purity 96%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Ammonium thiosulfate, Extra Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'