missing translation for 'onlineSavingsMsg'
Learn More

Amorolfine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7077754
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
78613-38-4
C21H35NO·HCl
353.97
XZKWIPVTHGWDCF-KUZYQSSXSA-N
amorolfine hydrochloride, amorolfine hcl, loceryl, curanail, locetar, odenil, amorolfin, pekiron, amorolfine hydrochloride jan
54259
CHEBI:59649
(2R,6S)-2,6-dimethyl-4-[2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl]morpholine;hydrochloride
CCC(C)(C)C1=CC=C(C=C1)CC(C)CN2CC(OC(C2)C)C.Cl
This item is not returnable. View return policy
78613-38-4
C21H35NO·HCl
353.97
XZKWIPVTHGWDCF-KUZYQSSXSA-N
amorolfine hydrochloride, amorolfine hcl, loceryl, curanail, locetar, odenil, amorolfin, pekiron, amorolfine hydrochloride jan
54259
CHEBI:59649
(2R,6S)-2,6-dimethyl-4-[2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl]morpholine;hydrochloride
CCC(C)(C)C1=CC=C(C=C1)CC(C)CN2CC(OC(C2)C)C.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 221.0°C to 223.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: 9g/L. Other solubilities: 28 mg/mL dmso, 71 mg/mL ethanol
Formula Weight 353.97
Percent Purity 98%
Chemical Name or Material Amorolfine hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Amorolfine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'