Learn More
Emma Safety Footwear Andes XD Safety Shoes
Features a steel toe cap and an integrated stainless steel plate. The double-layer, anti-slip PU sole provides extra protection from hazards on the ground.
124.02 € - 125.08 €
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15998321
|
Emma Safety Footwear
MM30556837 |
37 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15908331
|
Emma Safety Footwear
MM30556838 |
38 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15918331
|
Emma Safety Footwear
MM30556839 |
39 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15928331
|
Emma Safety Footwear
MM30556840 |
40 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15938331
|
Emma Safety Footwear
MM30556841 |
41 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15948331
|
Emma Safety Footwear
MM30556842 |
42 |
125.08 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15958331
|
Emma Safety Footwear
MM30556843 |
43 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15968331
|
Emma Safety Footwear
MM30556844 |
44 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15978331
|
Emma Safety Footwear
MM30556845 |
45 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15988331
|
Emma Safety Footwear
MM30556846 |
46 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15998331
|
Emma Safety Footwear
MM30556847 |
47 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15908341
|
Emma Safety Footwear
MM30556848 |
48 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
15918341
|
Emma Safety Footwear
MM30556849 |
49 |
124.02 €
1 pair |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Andes XD | |
Oiled Nubuck |