missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves

Catalog Number 15399622
164.82 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.


Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.

  • Provide flexibility and tactility
  • Made from an exceptionally thin chemical-resistant, five-layer laminated film
  • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
  • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
  • Hand specific
  • Broad spectrum chemical resistance
  • Wing-thumb for added wearer comfort

 

Certifications/Compliance (Europe) CE 0493, EN420:2003 + A1:2009; Category lll
Color White
Hand Specific
Latex Free Yes
Material Laminated Film
Size 11
Thickness (English) 2.5 mil
Thickness (Metric) 0.06 mm
EN388 Standard EN 388:0000
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABCEFGHIMS
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Gauntlet
Grip Design Smooth
Lining Unlined
Model 2-100 11 902214
Series AlphaTec™ 02-100
Length (English) 15 in.
Length (Metric) 38 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'