Learn More
14-3-3 sigma Monoclonal Antibody (1433S01), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511663
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-11663 targets 14-3-3 sigma in ICC/IF, IHC (P), IP, and WB applications and shows reactivity with Human samples. The MA5-11663 immunogen is recombinant human 1433 sigma protein.
14-3-3 sigma Monoclonal antibody specifically detects 14-3-3 sigma in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
14-3-3 sigma | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P31947 | |
SFN | |
Recombinant human 1433 sigma protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1433S01 | |
Unconjugated | |
SFN | |
14-3-3 protein sigma; 14-3-3 sigma; 14-3-3 sigma protein; 14-3-3sigma; epithelial cell marker protein 1; Er; HGNC:10773; HME1; Mkrn3; Mme1; SFN; Stratifin; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, sigma polypeptide; Ywhas | |
Mouse | |
Protein G | |
RUO | |
2810 | |
4° C | |
Liquid |