Learn More
6x-His Tag Monoclonal Antibody (HIS.H8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121315
249.80 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : LE
Additional Details : Weight : 1.24740kg
Description
MA1-21315 may be stored at 4° C short term. Add 0.05% sodium azide if desired. For long term storage, store at -20° C, avoiding freeze/thaw cycles.
6x-His Tag Monoclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
6x-His Tag | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
HIS.H8 | |
Unconjugated | |
6His tag; 6X His; His Tag; Histidine Tag | |
6x His synthetic peptide. | |
100 μg | |
Primary | |
Antibody | |
IgG2b |