missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6x-His Tag Monoclonal Antibody (HIS.H8), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121315BTIN
253.65 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA1-21315-BTIN has been successfully used for Western blot, ELISA, IF and IP
The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombit proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombit protein using genetic engineering. By choosing a particular epitope and recombit protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombit proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.Specifications
6x-His Tag | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
Mouse | |
Protein A | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
HIS.H8 | |
Biotin | |
His Tag; Histidine Tag; Histidine Epitope Tag; 6X His eptope tag; 6His tag; 6X His | |
6x His synthetic peptide. | |
50 μL | |
Primary | |
Antibody | |
IgG2b |