missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132357
Code : LE
Additional Details : Weight : 0.01000kg
Description
Store as a concentrated solution. Centrifuge briefly prior to opening vial.
ABCE1 Polyclonal antibody specifically detects ABCE1 in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
ABCE1 | |
Polyclonal | |
Unconjugated | |
ABCE1 | |
2'-5'-oligoadenylate-binding protein; ABC 38; ABC38; ABCE 1; ABCE1; ATP binding cassette subfamily E member 1; ATP-binding cassette sub-family E member 1; ATP-binding cassette, subfamily E (OABP), member 1; ATP-binding cassette, sub-family E (OABP), member 1; ATP-binding cassette, subfamily E, member 1; C79080; HP68; HuHP68; OABP; OK/SW-cl.40; Ribonuclease 4 inhibitor; ribonuclease L (2', 5'-oligoisoadenylate synthetase-dependent) inhibitor; ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent) inhibitor; RLI; RLI1; RNase 4 inhibitor; RNase L inhibitor; RNase L inhibitor 1; RNASEL1; Rnaseli; RNS41; RNS4I; RNS4l (Eye) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6059 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P61221 | |
ABCE1 | |
A fusion protein containing amino acids 1-119 of the ABCE1 protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |