Learn More
ABI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534783
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: PC-3, NIH-3T3, BCL-1, Raw264.7, C2C12. Predicted reactivity: Mouse (98%), Rat (96%), Chicken (85%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ABI1 Polyclonal antibody specifically detects ABI1 in Human, Mouse samples. It is validated for Western Blot.Specifications
ABI1 | |
Polyclonal | |
Unconjugated | |
ABI1 | |
abelson interactor 1; Abi1; Abi-1; Abl interactor 1; abl-binding protein 4; AblBP4; abl-interactor 1; Abl-interactor protein 1 long; Ablphilin-1; E3b1; EL2; eps8 binding protein; eps8 binding protein (e3B1), alternatively spliced; eps8 SH3 domain-binding protein; Eps8-binding protein; HPP; HS3; interactor protein AblBP4; LOW QUALITY PROTEIN: abl interactor 1; NAP1; nap1 binding protein; Nap1-binding protein; Nap1BP; spectrin SH3 domain binding protein 1; spectrin SH3 domain-binding protein 1; SPH3; SPTA; SSH3BP; Ssh3bp1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10006, 11308 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8CBW3, Q8IZP0 | |
ABI1 | |
Recombinant fragment contains a sequence corresponding to a region within amino acids 53 and 367 of SSH3BP1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |