missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA141700
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: Hela whole cell extract, HeLa, antigen standard for AKT3 (transient overexpression lysate).
AKT3 Polyclonal antibody specifically detects AKT3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
AKT3 | |
Polyclonal | |
Unconjugated | |
AKT3 | |
AI851531; AKT serine/threonine kinase 3; Akt3; AKT3 kinase; akt3a; D930002M15Rik; DKFZP434N0250; MPPH; MPPH2; Nmf350; PKB gamma; PKBG; PKB-GAMMA; PRKBG; Protein kinase Akt-3; protein kinase B gamma; protein kinase B, gamma; RAC-gamma; RAC-gamma serine/threonine protein kinase; RAC-gamma serine/threonine-protein kinase; RAC-PK-gamma; si:ch211-221n23.2; STK-2; thymoma viral proto-oncogene 3; v-akt murine thymoma viral oncogene 3; v-akt murine thymoma viral oncogene homolog 3; v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma); v-akt murine thymoma viral oncogene homolog 3a; v-akt murine thymoma viral oncogene-like 3 | |
Rabbit | |
Protein G | |
RUO | |
10000, 23797, 29414 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q63484, Q9WUA6, Q9Y243 | |
AKT3 | |
Synthetic peptide corresponding to residues 119-136 of human Akt3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |