missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alivin 1 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA146258
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody has 81% sequence homology with human.
A novel primary response gene, Alivin 1 (ali1), is an activity-dependent gene and promotes survival of neurons. Sequence analyses reveal that rat, mouse, and human Ali1 proteins contain seven leucine-rich repeats, one IgC2-like loop and a transmembrane domain, and display homology to Kek and Trk families. Results suggest that expression of ali1 promotes depolarization-dependent survival of the granule neuron. Mouse ali1 was mapped to a locus approximately 55. 3 cM from the centromere on chromosome 15 that is syntenic to positional candidate loci for familial Alzheimer′s disease type 5 and Parkinson′s disease 8 on human chromosome 12.Specifications
Alivin 1 | |
Polyclonal | |
Unconjugated | |
Q80ZD9, Q7TNJ4 | |
AMIGO2 | |
Synthetic peptide mapping to residues 449-464 of mouse and rat Alivin 1. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
Amigo2 | |
Amphoterin-induced protein 2, AMIGO-2, ALI1, DEGA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
105827, 300186 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |