missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p41-ARCb Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518047
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
p41-ARCb Polyclonal antibody specifically detects p41-ARCb in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
p41-ARCb | |
Polyclonal | |
Unconjugated | |
ARPC1B | |
41kDa; AA408064; AA408534; AA571392; actin related protein 2/3 complex subunit 1B; actin related protein 2/3 complex subunit 1B, 41kDa; actin related protein 2/3 complex, subunit 1B; actin related protein 2/3 complex, subunit 1B, 41kDa; actin-related protein 2/3 complex subunit 1B; Actin-related protein complex 1b; AF007010; ARC41; Arp2/3 complex 41 kDa subunit; ARP2/3 protein complex subunit p41; Arpc1b; AW208418; L72; OTTHUMP00000206020; p40-ARC; p41-ARC; SOP2Hs | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10095 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O15143 | |
ARPC1B | |
Synthetic peptide sequence (AYHSFLVEPISCH) corresponding to the N-terminus amino acids of ARPC1B | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |