Learn More
ATRIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1519
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-519 detects human and mouse ATRIP. PA1-519 has been successfully used in Western blot, immunoprecipitation and immunofluorescence procedures. By Western blot, this antibody detects an ∽ 86 kDa protein from human HeLa cells. PA1-519 antigen corresponds to amino acids 1-182 of human ATRIP
ATRIP Polyclonal antibody specifically detects ATRIP in Human, Mouse samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
ATRIP | |
Polyclonal | |
Unconjugated | |
ATRIP | |
6620401K05Rik; AGS1; ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR interacting protein; ATR IP; ATR-interacting protein; Atrip; DNase III; DRN3; FLJ12343; unnamed protein product | |
Rabbit | |
Protein G | |
RUO | |
235610, 84126 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.02% sodium azide | |
Q8BMG1, Q8WXE1 | |
ATRIP | |
Immunogen corresponds to amino acids 1-182 of human ATRIP. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |