missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA511379
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine based on sequence homology.
Bcl-2 Polyclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Bcl-2 | |
Polyclonal | |
Unconjugated | |
Bcl2 | |
apoptosis regulator Bcl-2; AW986256; B cell leukemia/lymphoma 2; B cell lymphoma 2 associated oncogene; B-cell CLL/lymphoma 2; B-cell leukemia/lymphoma 2; Bcl 2; BCL2; bcl-2; BCL2 apoptosis regulator; Bcl-2 protein; BCL2, apoptosis regulator; Bcl2-like protein; C430015F12Rik; D630044D05Rik; D830018M01Rik; OTTHUMP00000163680; PPP1R50; protein phosphatase 1, regulatory subunit 50 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
596 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2.0 mg/mL | |
PBS with 0.09% sodium azide | |
P10415 | |
Bcl2 | |
KLH conjugated synthetic peptide between 75-110 amino acids from human Bcl | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |