missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-2 Monoclonal Antibody (100221), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523763
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In Western blots, this antibody shows less than 1% cross-reactivity with recombinant human (rh) BMP-3, rhBMP-4, rhBMP-5, rhBMP-6, or rhBMP-7. Reconstitute at 0.5 mg/mL in sterile PBS.
BMP-2 Monoclonal antibody specifically detects BMP-2 in Human samples. It is validated for ELISA, Western Blot.Specifications
BMP-2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P12643 | |
BMP2 | |
CHO-derived recombinant human BMP-2 Gln283-Arg396 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Western Blot | |
100221 | |
Unconjugated | |
BMP2 | |
2610024H22Rik; AI467020; AL117858; AW546137; BB189135; BDA2; BMP; BMP2; BMP-2; BMP2A; BMP-2A; BMPRII; BMPR-II; Bone morphogenetic protein; Bone morphogenetic protein 2; bone morphogenetic protein 2 precursor (BMP-2) (BMP-2A); bone morphogenetic protein 2A; H-BMP-2; morphogen | |
Mouse | |
Protein A/G | |
RUO | |
650 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |