Learn More
CEA Monoclonal Antibody (1106), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514675
Code : LE
Additional Details : Weight : 0.01000kg
Description
The predicted MW of CEA is ∽77kD, but by Western blot MA5-14675 detects CEA with varying degrees of glycosylation at ∽77-180kD. Product MA514675 is a smaller package size of MIC0102 (formerly sold as a Seradyn product).
CEA Monoclonal antibody specifically detects CEA in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot.Specifications
CEA | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P06731 | |
Ceacam5 | |
Purified CEA from pooled tumors | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
1106 | |
Unconjugated | |
Ceacam5 | |
1600029H12Rik; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 5; CD66e; CEA; CEACAM5; Meconium antigen 100; OTTHUMP00000199033; pregnancy-specific glycoprotein 30; Psg30; sCD66e; soluble CD66e | |
Mouse | |
Protein A | |
RUO | |
1048 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |