missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11a Monoclonal Antibody (M17/4), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA110077
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD11a Monoclonal antibody specifically detects CD11a in Mouse samples. It is validated for Flow Cytometry.Specifications
CD11a | |
Monoclonal | |
0.5 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P24063 | |
ITGAL | |
C57BL/6 mouse splenic secondary cytotoxic T lymphocytes | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
M17/4 | |
FITC | |
ITGAL | |
(p180); alpha L integrin; antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; antigen CD11A p180; CD11 antigen-like family member A; CD11a; integrin alpha L; integrin alphaL; integrin alpha-L; integrin alpha-L precursor; integrin gene promoter; integrin subunit alpha L; integrin surface receptor; integrin, alpha L; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); ITGAL; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte function-associated molecule 1 alpha chain; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; Ly-15; Ly-21; lymphocyte antigen 15; lymphocyte function associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen-1 | |
Rat | |
Size-exclusion chromatography | |
RUO | |
16408 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |