Learn More
CD90 Mouse anti-Human, PE, Clone: 5E10, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555596
Additional Details : Weight : 0.01000kg
Description
Reacts with a 25-35kDa molecule expressed on 1-4% of human fetal liver cells, cord blood cells, and bone marrow cells. Anti-CD90 reacts with a subset of immature, CD34+ celsl and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is highly enriched for cells capable of long-term culture. Anti-CD90 is useful for enriching high proliferative potential colony-forming cells (HIPP-CFC) which are primative progenitor cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Specifications
CD90 | |
Monoclonal | |
0.2mg/mL | |
THY1; Thy-1 antigen; Thy-1 membrane glycoprotein | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
5.00E+10 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human HEL Cell Line | |
0.1 mg | |
Immunology | |
Baboon, Canine, Cynomolgus, Human, Porcine, Rhesus Monkey | |
IgG1 κ |
For Research Use Only.