Learn More
Chlamydia trachomatis MOMP Polyclonal Antibody, FITC, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA173073
Code : LD
Additional Details : Weight : 0.01000kg
Description
The PA1-73073 antibody reacts with Chlamydia trachomatis MOMP. Does not react with C. psittacii or C. pneumoniae in MIF. PA1-73073 has been successfully used in Immunofluorescence applications. The immunogen for PA1-73073 is purified MOMP from strain L2.
Chlamydia trachomatis MOMP Polyclonal antibody specifically detects Chlamydia trachomatis MOMP in Bacteria samples. It is validated for Immunocytochemistry.Specifications
Chlamydia trachomatis MOMP | |
Polyclonal | |
FITC | |
C. trachomatis MOMP; Chlamydia MOMP | |
Purified MOMP from strain L2. | |
1 mL | |
Primary | |
-20° C, store in dark | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide | |
Goat | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |