missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL12A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538890
Code : NEW
Additional Details : Weight : 0.00900kg
Description
This gene encodes the alpha chain of type XII collagen, a member of the FACIT (fibril-associated collagens with interrupted triple helices) collagen family. Type XII collagen is a homotrimer found in association with type I collagen, an association that is thought to modify the interactions between collagen I fibrils and the surrounding matrix. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008].Specifications
COL12A1 | |
Polyclonal | |
Unconjugated | |
COL12A1 | |
COL12A1 | |
A synthetic peptide derived from the internal region of human Collagen XII alpha1 | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q99715 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1303 | |
-20°C | |
Liquid |