missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4 Mouse anti-Human, Clone: 49H9, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0188
Additional Details : Weight : 0.99000kg
Description
A suggested positive control for this product is human plasma.
This gene encodes the acidic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus and type I diabetes mellitus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene.Specifications
Complement C4 | |
Monoclonal | |
Unconjugated | |
P0C0L4 | |
C4A | |
Protein purified from Human plasma | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Immunoprecipitation, Western Blot | |
49H9 | |
C4A | |
DAMC-258G8.6, C4, C4A2, C4A3, C4A4, C4A6, C4S, CO4, CPAMD2, MGC164979, RG | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
720 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |