missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 2e Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531023
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cytokeratin 2e Polyclonal antibody specifically detects Cytokeratin 2e in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Cytokeratin 2e | |
Polyclonal | |
Unconjugated | |
KRT2 | |
BB005427; CK 2e; CK-2e; Cytokeratin-2e; epithelial keratin-2e; K2e; Kb2; keratin 2; keratin 2 epidermis; keratin 2, type II; keratin 2e; keratin complex 2, basic, gene 17; keratin complex 2, gene 17; keratin, type II cytoskeletal 2 epidermal; Keratin-2 epidermis; keratin-2e; KRT2; krt2 {ECO:0000250; Krt2-17; Krt2-2; KRT2A; Krt2e; KRTE; MGC116967; MGC116968; type II keratin Kb2; type-II keratin Kb2; UniProtKB:P35908} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3849 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.95 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P35908 | |
KRT2 | |
Recombinant protein fragment corresponding to a region within amino acids 18 and 205 of Human Cytokeratin 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |