Learn More
DDX54 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541295
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GPNRGAKRRR EEARQRDQEF YIPYRPKDFD SERGLSISGE GGAFEQQAAG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 92%; Rat: 93%; Zebrafish: 92%
DDX54 Polyclonal antibody specifically detects DDX54 in Human samples. It is validated for Western Blot.Specifications
DDX54 | |
Polyclonal | |
Unconjugated | |
DDX54 | |
2410015A15Rik; 5-Apr; AI414901; ATP-dependent RNA helicase; ATP-dependent RNA helicase DDX54; ATP-dependent RNA helicase DP97; chunp6913; Ddx54; DEAD (Asp-Glu-Ala-Asp) box polypeptide 54; DEAD box helicase 97 KDa; DEAD box protein 54; DEAD box RNA helicase 97 kDa; DEAD-box helicase 54; DP97; LOW QUALITY PROTEIN: ATP-dependent RNA helicase DDX54; mgc2835; RGD1562539; zgc:111908 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79039 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TDD1 | |
DDX54 | |
Synthetic peptide directed towards the C-terminal of human DDX54 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |