Learn More
DOK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532384
Code : A0
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast carcinoma.
DOK1 Polyclonal antibody specifically detects DOK1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
DOK1 | |
Polyclonal | |
PBS with 1% BSA and <0.1% sodium azide; pH 7.6 | |
Q99704 | |
DOK1 | |
Synthetic peptide corresponding to N-terminus of human DOK-1 protein. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
DOK1 | |
AW557123; Docking protein 1; docking protein 1, 62kDa (downstream of tyrosine kinase 1); Dok; DOK 1; DOK1; Dok-1; Downstream of tyrosine kinase 1; downstream of tyrosine kinase-1; MGC117395; MGC138860; p240; p62 dok; p62(dok); p62DOK; pp62; TLP1; TP1; TROVE domain family; TROVE1; VAULT2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1796 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |