Learn More
Dystrophin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA137587
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, chicken, canine and porcine based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is skeletal muscle tissue.
Dystrophin Polyclonal antibody specifically detects Dystrophin in Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Dystrophin | |
Polyclonal | |
Unconjugated | |
DMD | |
apodystrophin-3; apodystrophin-I; BMD; CMD3B; dmd; dmd.1; dmd.1.L; DNADMD1; Dp427; Dp71; Duchenne muscular dystrophy (DMD); DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; DXSmh7; DXSmh9; dys; Dystrophin; dystrophin Dp40; dystrophin Dp71 isoform; dystrophin Dp71a; dystrophin Dp71ab; dystrophin Dp71b; dystrophin, gene 1 L homeolog; dystrophin, muscular dystrophy; GS1-19O24.1; mdx; MRX85; Muscular dystrophy Duchenne and Becker types; OTTHUMP00000215592; pke; utrn; utrophin; XELAEV_18012903mg; X-linked muscular dystrophy | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13405, 1756 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Conc. not determined | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P11531, P11532 | |
DMD | |
Synthetic peptide derived from C-terminus of human dystrophin. | |
1 mL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |