missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129595
Code : LE
Additional Details : Weight : 0.01000kg
Description
Expected to cross react with chicken (100% identity), human (100% identity), mouse, (94% identity), Xenopus laevis (100% identity) due to sequence homology.
GLI3 Polyclonal antibody specifically detects GLI3 in Avian, Canine, Chicken, Chimpanzee, Human, Mouse, Non-human Primate, X. laevis, Zebrafish samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
GLI3 | |
Polyclonal | |
Unconjugated | |
GLI3 | |
ACLS; add; AI854843; anterior digit pattern deformity; AU023367; Bph; brachyphalangy; extra toes; GCPS; GLI family zinc finger 3; GLI family zinc finger 3 L homeolog; gli3; GLI3 C-terminally truncated form; GLI3 form of 190 kDa; GLI3 form of 83 kDa; GLI3 full length protein; GLI3 full-length protein; gli3.L; GLI3-190; GLI3-83; GLI3FL; GLI-Kruppel family member 3; GLI-Kruppel family member GLI3; GLI-Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome); glioma-associated oncogene family zinc finger 3; im:7142603; neural specific DNA binding protein; neural-specific DNA-binding protein xGLI3; oncogene GLI3; PAPA; PAP-A; PAPA1; PAPB; Pdn; PHS; polydactyly Nagoya; PPDIV; Transcriptional activator GLI3; Transcriptional repressor GLI3R; XELAEV_18031191mg; xgli3; xGLI-3; Xt; Zinc finger protein GLI3; zinc finger protein GLI3; transcriptional activator GLI3; zinc finger transcription factor Gli3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14634, 2737, 397837, 403042, 420769, 463369, 483244 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
P10071, Q5IS56, Q61602, Q91660, Q9IA31 | |
GLI3, gli3.L | |
synthetic peptide NEDESPGQTYHRERRNA-C conjugated to KLH, corresponding to amino acids 41-57 of Human GLI3. | |
50 μg | |
Primary | |
Avian, Canine, Chicken, Chimpanzee, Human, Mouse, Non-human Primate, X. laevis, Zebrafish | |
Antibody | |
IgG |