Learn More
Goat IgG Isotype Control, Invitrogen™
Goat Isotype Control
Brand: Invitrogen 026202
95.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Goat IgG is purified from pooled normal goat serum using various methods. Purity is verified by SDS-PAGE analysis. The concentration of 5 mg/mL based on an E (1%) = 14.0 at 280 nm.
Goat IgG, also known as goat immunoglobulin G, is an antibody that is produced by the immune system of goats in response to the presence of antigens. IgG is a type of immunoglobulin that plays a role in the recognition and neutralization of pathogens, such as bacteria and viruses. It binds to specific antigens and triggers various immune responses, including phagocytosis and complement activation. Goat IgG can be used in research to study the immune response to pathogens, as well as to develop diagnostic tests and therapeutic interventions for infectious diseases.Specifications
Goat IgG | |
5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Goat | |
purified | |
RUO | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
IgG |
Control | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG | |
Gamma Immunoglobin | |
10 mg | |
Not Applicable | |
Liquid |